siRNA Details
| Gene Symbol: ADO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032804.5 | NP_116193.2 | 1 | 2792 | ||
| GenBank mRNA | BC028589.2 | - | 1 | 2662 | 4838951 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADO| Gene Symbol: ADO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032804.5 | NP_116193.2 | 1 | 2792 | ||
| GenBank mRNA | BC028589.2 | - | 1 | 2662 | 4838951 | |