siRNA Details
| Gene Symbol: LTV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032860.3 | NP_116249.2 | 3 | 355 | ||
| GenBank mRNA | AK027815.1 | - | 1 | 104 | ||
| AK095269.1 | - | 2 | 1818 | |||
| AK300360.1 | - | 3 | 341 | |||
| BC009855.2 | - | 3 | 355 | 3926227 | ||

LTV1
Genomic Map