siRNA Details
| Gene Symbol: LTV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032860.3 | NP_116249.2 | 9, 10 | 1231 | ||
| GenBank mRNA | AK027815.1 | - | 7, 8 | 980 | ||
| AK091447.1 | - | 4, 5 | 4280 | |||
| BC009855.2 | - | 9, 10 | 1231 | 3926227 | ||

LTV1
Genomic Map