siRNA Details
| Gene Symbol: SERAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032861.3 | NP_116250.3 | 7 | 664 | ||
| NR_073096.1 | 7 | 664 | ||||
| XM_006715586.2 | XP_006715649.1 | 6 | 593 | |||
| XM_011536196.2 | XP_011534498.1 | 6 | 687 | |||
| XM_011536198.2 | XP_011534500.1 | 5 | 589 | |||
| GenBank mRNA | AK027823.1 | - | 7 | 654 | ||
| AK125878.1 | - | 8 | 1856 | |||
| BC001705.1 | - | 7 | 621 | 3350731 | ||
| BX648104.1 | - | 6 | 596 | |||
| DB046171.1 | - | 6 | 538 | |||

SERAC1
Genomic Map