siRNA Details
| Gene Symbol: SERAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032861.3 | NP_116250.3 | 17 | 2046 | ||
| XM_006715586.2 | XP_006715649.1 | 16 | 1975 | |||
| XM_011536196.2 | XP_011534498.1 | 16 | 2069 | |||
| XM_011536198.2 | XP_011534500.1 | 15 | 1971 | |||
| GenBank mRNA | AK023656.1 | - | 1 | 82 | ||
| AK027823.1 | - | 17 | 2036 | |||
| AK125878.1 | - | 18 | 3238 | |||
| BC001705.1 | - | 17 | 2003 | 3350731 | ||
| BX648104.1 | - | 15 | 3422 | |||

SERAC1
Genomic Map