siRNA Details
| Gene Symbol: MFSD14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033055.2 | NP_149044.2 | 12 | 1684 | ||
| XM_005271118.1 | XP_005271175.1 | 13 | 1960 | |||
| XM_011541957.1 | XP_011540259.1 | 11 | 1725 | |||
| XM_017002084.1 | XP_016857573.1 | 13 | 1927 | |||
| GenBank mRNA | AK057172.1 | - | 12 | 1820 | ||
| AK075208.1 | - | 6 | 885 | |||
| AK096669.1 | - | 9 | 1335 | |||
| AK127671.1 | - | 1 | 1196 | |||
| BC064409.1 | - | 9 | 1341 | |||
| BC107768.1 | - | 9 | 1433 | |||

MFSD14A
Genomic Map