siRNA Details
| Gene Symbol: RRP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033112.2 | NP_149103.1 | 7 | 1092 | ||
| GenBank mRNA | AF161371.1 | - | 6 | 847 | ||
| BC006293.2 | - | 7 | 1086 | |||
| BC011933.2 | - | 7 | 1092 | 4300313 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRP36| Gene Symbol: RRP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033112.2 | NP_149103.1 | 7 | 1092 | ||
| GenBank mRNA | AF161371.1 | - | 6 | 847 | ||
| BC006293.2 | - | 7 | 1086 | |||
| BC011933.2 | - | 7 | 1092 | 4300313 | ||