siRNA Details
| Gene Symbol: PLCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033123.3 | NP_149114.2 | 4, 5 | 628 | ||
| NR_073075.1 | 4, 5 | 631 | ||||
| XM_005253519.2 | XP_005253576.1 | 4, 5 | 621 | |||
| XM_017020176.1 | XP_016875665.1 | 4, 5 | 621 | |||
| XM_017020177.1 | XP_016875666.1 | 4, 5 | 621 | |||
| XM_017020178.1 | XP_016875667.1 | 4, 5 | 621 | |||
| XM_017020179.1 | XP_016875668.1 | 4, 5 | 616 | |||
| XM_017020180.1 | XP_016875669.1 | 4, 5 | 510 | |||
| XM_017020181.1 | XP_016875670.1 | 4, 5 | 575 | |||
| GenBank mRNA | AF532185.1 | - | 4, 5 | 553 | ||
| AK292279.1 | - | 4, 5 | 626 | |||
| AK314516.1 | - | 4, 5 | 584 | |||
| AY035866.1 | - | 4, 5 | 590 | |||

PLCZ1
Genomic Map