siRNA Details
| Gene Symbol: OR51B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033180.4 | NP_149420.4 | 1 | 287 | ||
| GenBank mRNA | BC117461.1 | - | 1 | 287 | 40126012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR51B2| Gene Symbol: OR51B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033180.4 | NP_149420.4 | 1 | 287 | ||
| GenBank mRNA | BC117461.1 | - | 1 | 287 | 40126012 | |