siRNA Details
| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF333704.2 | - | 8, 9 | 1479 | ||
| AY017307.1 | - | 8, 9 | 1374 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSMD1| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF333704.2 | - | 8, 9 | 1479 | ||
| AY017307.1 | - | 8, 9 | 1374 | |||