siRNA Details
| Gene Symbol: TNKS1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033396.2 | NP_203754.2 | 5 | 1088 | ||
| XM_006718725.3 | XP_006718788.1 | 5 | 1104 | |||
| XM_011545325.2 | XP_011543627.1 | 5 | 1101 | |||
| GenBank mRNA | AB051528.2 | - | 5 | 1068 | ||
| AF441771.1 | - | 4 | 1230 | |||
| AK124903.1 | - | 5 | 1115 | |||
| AK127871.1 | - | 3 | 511 | |||
| BC150333.1 | - | 5 | 1079 | 8860508 | ||

TNKS1BP1
Genomic Map