siRNA Details
| Gene Symbol: ITPRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272012.1 | NP_001258941.1 | 2 | 571 | ||
| NM_001272013.1 | NP_001258942.1 | 2 | 691 | |||
| NM_033397.3 | NP_203755.1 | 3 | 883 | |||
| XM_005270257.2 | XP_005270314.1 | 1 | 447 | |||
| GenBank mRNA | AB051541.1 | - | 2 | 602 | ||
| AK056526.1 | - | 2 | 503 | |||
| AK129877.1 | - | 3 | 594 | |||
| BC040536.1 | - | 2 | 543 | 5265379 | ||
| BC070108.1 | - | 3 | 883 | 4837204 | ||
| BX436559.2 | - | 2 | 570 | |||

ITPRIP
Genomic Map