siRNA Details
| Gene Symbol: SLC38A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033518.3 | NP_277053.2 | 4 | 297 | ||
| XM_005272694.3 | XP_005272751.2 | 3 | 287 | |||
| XM_005272695.4 | XP_005272752.3 | 3 | 289 | |||
| XM_005272697.2 | XP_005272754.2 | 3 | 240 | |||
| XM_005272698.4 | XP_005272755.2 | 3 | 178 | |||
| XM_006724569.3 | XP_006724632.1 | 4 | 297 | |||
| XM_017029960.1 | XP_016885449.1 | 3 | 209 | |||
| XM_017029961.1 | XP_016885450.1 | 3 | 181 | |||
| GenBank mRNA | AF276889.1 | - | 3 | 962 | ||
| AK094788.1 | - | 3 | 216 | |||
| BC019246.2 | - | 3 | 147 | 3506044 | ||
| DA089769.1 | - | 4 | 298 | |||

SLC38A5
Genomic Map