siRNA Details
| Gene Symbol: HIST1H2BK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080593.2 | NP_542160.1 | 2 | 562 | ||
| GenBank mRNA | AJ223352.1 | - | 2 | 518 | ||
| BC000893.1 | - | 2 | 544 | 3463930 | ||
| BC051872.1 | - | 2 | 542 | 6526835 | ||
| BC064959.1 | - | 2 | 542 | 6093977 | ||
| BC108737.1 | - | 2 | 542 | 4707078 | ||
| DA873786.1 | - | 2 | 562 | |||

HIST1H2BK
Genomic Map