siRNA Details
| Gene Symbol: ASB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080863.4 | NP_543139.4 | 5 | 2057 | ||
| GenBank mRNA | AK054727.1 | - | 5 | 2057 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASB16| Gene Symbol: ASB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080863.4 | NP_543139.4 | 5 | 2057 | ||
| GenBank mRNA | AK054727.1 | - | 5 | 2057 | ||