siRNA Details
| Gene Symbol: TMEM183A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322955.1 | NP_001309884.1 | 4, 5 | 619 | ||
| NM_001322956.1 | NP_001309885.1 | 4, 5 | 619 | |||
| NM_001322957.1 | NP_001309886.1 | 4, 5 | 622 | |||
| NM_001322958.1 | NP_001309887.1 | 3, 4 | 532 | |||
| NM_001322959.1 | NP_001309888.1 | 3, 4 | 529 | |||
| NM_138391.4 | NP_612400.3 | 4, 5 | 583 | |||
| NR_136530.1 | 4, 5 | 622 | ||||
| NR_136531.1 | 4, 5 | 619 | ||||
| NR_136532.1 | 4, 5 | 622 | ||||
| NR_136533.1 | 4, 5 | 622 | ||||
| NR_136534.1 | 4, 5 | 619 | ||||
| GenBank mRNA | AF070537.1 | - | 4, 5 | 600 | ||
| AK291426.1 | - | 4, 5 | 597 | |||
| AK309745.1 | - | 4, 5 | 594 | |||
| AK312985.1 | - | 4, 5 | 597 | |||
| BC013073.1 | - | 1 | 583 | 4151716 | ||
| BC038952.2 | - | 1 | 635 | 5740484 | ||
| BC067775.1 | - | 4, 5 | 593 | 4817183 | ||
| BC150647.1 | - | 4, 5 | 565 | 9057018 | ||
| Gene Symbol: TMEM183B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079809.1 | NP_001073277.1 | 1 | 581 | ||
| GenBank mRNA | AK297222.1 | - | 4, 5 | 501 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_020588, NM_001042485, AB041592, AK004922, AK152852, and others.

TMEM183A
Genomic Map