siRNA Details
| Gene Symbol: MCFD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171506.2 | NP_001164977.1 | 5 | 2778 | ||
| NM_001171507.2 | NP_001164978.1 | 4 | 2670 | |||
| NM_001171508.2 | NP_001164979.1 | 4 | 2822 | |||
| NM_001171509.2 | NP_001164980.1 | 3 | 2529 | |||
| NM_001171510.2 | NP_001164981.1 | 3 | 2515 | |||
| NM_001171511.2 | NP_001164982.1 | 3 | 2667 | |||
| NM_139279.5 | NP_644808.1 | 4 | 2684 | |||
| GenBank mRNA | AF537214.1 | - | 4 | 2621 | ||
| AK095006.1 | - | 1 | 1391 | |||
| AL833900.1 | - | 4 | 2619 | |||
| BC040357.2 | - | 4 | 2610 | 3925166 | ||
| CR749562.1 | - | 3 | 2472 | |||

MCFD2
Genomic Map