siRNA Details
Gene Symbol: HN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144570.2 | NP_653171.1 | 5 | 3175 | ||
GenBank mRNA | BC060853.1 | - | 5 | 3169 | 5296086 | |
BM474060.1 | - | 1 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144570.2 | NP_653171.1 | 5 | 3175 | ||
GenBank mRNA | BC060853.1 | - | 5 | 3169 | 5296086 | |
BM474060.1 | - | 1 | 88 |