siRNA Details
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113525.1 | NP_001106997.1 | 7 | 1324 | ||
| NM_152287.3 | NP_689500.2 | 7 | 1314 | |||
| NR_110122.1 | 7 | 1314 | ||||
| NR_110126.1 | 7 | 1282 | ||||
| NR_110128.1 | 7 | 1205 | ||||
| NR_110129.1 | 7 | 1294 | ||||
| XM_005256324.3 | XP_005256381.1 | 7 | 1273 | |||
| XM_005256328.3 | XP_005256385.1 | 5 | 954 | |||
| XM_017023890.1 | XP_016879379.1 | 6 | 1249 | |||
| GenBank mRNA | AK026482.1 | - | 7 | 1264 | ||
| AK056405.1 | - | 7 | 1265 | |||
| AK096762.1 | - | 1 | 1834 | |||
| AK289801.1 | - | 7 | 1173 | |||
| AK295770.1 | - | 5 | 986 | |||
| AK315793.1 | - | 4 | 681 | |||
| BC032781.1 | - | 6 | 983 | |||
| BC105959.1 | - | 7 | 1107 | 40036181 | ||
| BC107603.1 | - | 7 | 1139 | 40036180 | ||
| BC110348.1 | - | 7 | 1039 | 5196957 | ||

ZNF276
Genomic Map