siRNA Details
| Gene Symbol: PANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324191.1 | NP_001311120.1 | 3 | 1083 | ||
| NM_001324192.1 | NP_001311121.1 | 2 | 915 | |||
| NM_001324193.1 | NP_001311122.1 | 2 | 251 | |||
| NM_024960.5 | NP_079236.3 | 2 | 186 | |||
| NM_153638.3 | NP_705902.2 | 2 | 915 | |||
| NM_153640.3 | NP_705904.1 | 2 | 251 | |||
| NR_136715.1 | 2 | 915 | ||||
| XM_005260835.3 | XP_005260892.1 | 2 | 368 | |||
| XM_005260836.4 | XP_005260893.3 | 2 | 188 | |||
| XM_011529364.2 | XP_011527666.1 | 2 | 771 | |||
| XM_011529365.2 | XP_011527667.1 | 2 | 771 | |||
| XM_017028077.1 | XP_016883566.1 | 2 | 288 | |||
| XM_017028078.1 | XP_016883567.1 | 2 | 1629 | |||
| XM_017028079.1 | XP_016883568.1 | 2 | 199 | |||
| GenBank mRNA | AK021791.1 | - | 2 | 186 | ||
| AK097796.1 | - | 2 | 434 | |||
| AK310190.1 | - | 3 | 623 | |||
| AL519408.3 | - | 2 | 203 | |||
| AL532307.3 | - | 2 | 259 | |||
| BC009421.1 | - | 2 | 431 | |||
| BC065019.1 | - | 2 | 197 | |||
| BC107724.1 | - | 2 | 238 | |||
| BF698882.1 | - | 235 | ||||

PANK2
Genomic Map