siRNA Details
| Gene Symbol: PANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324191.1 | NP_001311120.1 | 8 | 2428 | ||
| NM_001324193.1 | NP_001311122.1 | 7 | 1453 | |||
| NM_024960.5 | NP_079236.3 | 7 | 1531 | |||
| NM_153638.3 | NP_705902.2 | 7 | 2260 | |||
| NM_153640.3 | NP_705904.1 | 7 | 1596 | |||
| NR_136715.1 | 7 | 2117 | ||||
| XM_005260835.3 | XP_005260892.1 | 7 | 1713 | |||
| XM_005260836.4 | XP_005260893.3 | 7 | 1533 | |||
| XM_011529364.2 | XP_011527666.1 | 6 | 1939 | |||
| XM_017028077.1 | XP_016883566.1 | 7 | 1490 | |||
| XM_017028078.1 | XP_016883567.1 | 7 | 2831 | |||
| XM_017028079.1 | XP_016883568.1 | 7 | 1401 | |||
| GenBank mRNA | AK021791.1 | - | 7 | 1531 | ||
| AK097796.1 | - | 7 | 1779 | |||
| AL713654.1 | - | 5 | 879 | |||
| BC009421.1 | - | 7 | 1633 | |||

PANK2
Genomic Map