siRNA Details
| Gene Symbol: PKHD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177531.4 | NP_803875.2 | 5, 6 | 587 | ||
| XM_011517371.2 | XP_011515673.1 | 5, 6 | 538 | |||
| XM_017013969.1 | XP_016869458.1 | 5, 6 | 538 | |||
| XM_017013970.1 | XP_016869459.1 | 5, 6 | 538 | |||
| XM_017013971.1 | XP_016869460.1 | 5, 6 | 538 | |||
| XM_017013972.1 | XP_016869461.1 | 5, 6 | 538 | |||
| GenBank mRNA | AY219181.1 | - | 5, 6 | 587 | ||
| BC093096.1 | - | 5, 6 | 558 | |||

PKHD1L1
Genomic Map