siRNA Details
| Gene Symbol: NBEAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114132.1 | NP_001107604.1 | 27 | 4337 | ||
| XM_005246787.3 | XP_005246844.1 | 27 | 4363 | |||
| XM_005246788.2 | XP_005246845.1 | 6 | 839 | |||
| XM_006712698.3 | XP_006712761.1 | 28 | 4450 | |||
| XM_006712699.3 | XP_006712762.1 | 28 | 4450 | |||
| XM_011511658.2 | XP_011509960.1 | 28 | 4405 | |||
| XM_011511659.2 | XP_011509961.1 | 28 | 4462 | |||
| XM_011511660.2 | XP_011509962.1 | 24 | 3784 | |||
| XM_011511662.2 | XP_011509964.1 | 20 | 3119 | |||
| XM_011511663.2 | XP_011509965.1 | 28 | 4440 | |||
| XM_017004728.1 | XP_016860217.1 | 15 | 2517 | |||
| XM_017004729.1 | XP_016860218.1 | 3 | 531 | |||
| GenBank mRNA | AK127772.1 | - | 1 | 205 | ||

NBEAL1
Genomic Map