siRNA Details
| Gene Symbol: KIRREL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032123.6 | NP_115499.5 | 15 | 1877 | ||
| NM_199179.3 | NP_954648.3 | 14 | 1727 | |||
| NM_199180.3 | NP_954649.3 | 15 | 2019 | |||
| XM_011527362.1 | XP_011525664.1 | 16 | 2079 | |||
| XM_011527363.1 | XP_011525665.1 | 16 | 2060 | |||
| XM_011527365.1 | XP_011525667.1 | 15 | 1929 | |||
| XM_011527366.1 | XP_011525668.1 | 14 | 2190 | |||
| XM_017027348.1 | XP_016882837.1 | 14 | 2481 | |||
| GenBank mRNA | AB593116.1 | - | 15 | 1983 | ||
| AK307865.1 | - | 14 | 2235 | |||
| AL136654.1 | - | 15 | 2003 | |||
| AY305301.1 | - | 15 | 1821 | |||
| AY305302.1 | - | 14 | 1671 | |||
| AY358742.1 | - | 15 | 2005 | |||
| BC007312.1 | - | 6 | 552 | |||
| BC064925.1 | - | 18 | 1942 | 6141879 | ||

KIRREL2
Genomic Map