siRNA Details
| Gene Symbol: TRIM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017583.5 | NP_060053.2 | 1 | 911 | ||
| XM_006718254.1 | XP_006718317.1 | 1 | 971 | |||
| GenBank mRNA | AJ249128.1 | - | 1 | 728 | ||
| AK021823.1 | - | 1 | 17 | |||
| AK074242.1 | - | 1 | 735 | |||
| BC013166.1 | - | 1 | 858 | 3924490 | ||
| BC024031.2 | - | 1 | 758 | 3532065 | ||

TRIM44
Genomic Map