siRNA Details
| Gene Symbol: TRIM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017583.5 | NP_060053.2 | 2 | 1061 | ||
| XM_006718254.1 | XP_006718317.1 | 2 | 1121 | |||
| GenBank mRNA | AK021823.1 | - | 2 | 167 | ||
| AK074242.1 | - | 2 | 885 | |||
| BC013166.1 | - | 2 | 1008 | 3924490 | ||
| BC024031.2 | - | 2 | 908 | 3532065 | ||

TRIM44
Genomic Map