siRNA Details
| Gene Symbol: KCTD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017634.3 | NP_060104.2 | 12 | 2280 | ||
| GenBank mRNA | AK000045.1 | - | 7 | 1608 | ||
| AK025995.1 | - | 1 | 326 | |||
| AL117436.1 | - | 1 | 3558 | |||
| BC068518.1 | - | 12 | 2094 | 5265435 | ||

KCTD9
Genomic Map