siRNA Details
| Gene Symbol: CNNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017649.4 | NP_060119.3 | 1 | 416 | ||
| NM_199076.2 | NP_951058.1 | 1 | 416 | |||
| NM_199077.2 | NP_951059.1 | 1 | 416 | |||
| XM_005269933.4 | XP_005269990.1 | 1 | 441 | |||
| XM_006717908.2 | XP_006717971.1 | 1 | 441 | |||
| XM_011539911.2 | XP_011538213.1 | 1 | 441 | |||
| GenBank mRNA | AK023479.1 | - | 1 | 377 | ||
| BC021222.2 | - | 1 | 416 | 4308662 | ||

CNNM2
Genomic Map