siRNA Details
| Gene Symbol: FERMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017671.4 | NP_060141.3 | 7, 8 | 1746 | ||
| GenBank mRNA | AB105105.1 | - | 7, 8 | 1746 | ||
| AF443278.1 | - | 7, 8 | 1315 | |||
| AK025365.1 | - | 2, 3 | 127 | |||
| AK092195.1 | - | 5, 6 | 804 | |||
| AK314394.1 | - | 7, 8 | 1136 | |||
| AY137240.1 | - | 6, 7 | 957 | |||
| BC035882.1 | - | 7, 8 | 1429 | 4941567 | ||
| BC040545.1 | - | 6, 7 | 1001 | 5277060 | ||
| BX647465.1 | - | 2, 3 | 153 | |||

FERMT1
Genomic Map