siRNA Details
| Gene Symbol: ZDHHC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145548.1 | NP_001139020.1 | 9 | 1231 | ||
| NM_017740.2 | NP_060210.2 | 8 | 1120 | |||
| XM_017023434.1 | XP_016878923.1 | 6 | 656 | |||
| XM_017023435.1 | XP_016878924.1 | 6 | 516 | |||
| XM_017023436.1 | XP_016878925.1 | 6 | 631 | |||
| GenBank mRNA | AK000286.1 | - | 8 | 1120 | ||
| AK001654.1 | - | 6 | 506 | |||
| AK225768.1 | - | 8 | 1072 | |||
| BC017702.1 | - | 6 | 481 | 4394921 | ||
| BC018772.1 | - | 9 | 1218 | 4869114 | ||

ZDHHC7
Genomic Map