siRNA Details
| Gene Symbol: G2E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308097.1 | NP_001295026.1 | 13 | 1912 | ||
| NM_017769.4 | NP_060239.2 | 14 | 1953 | |||
| XM_011536925.2 | XP_011535227.1 | 14 | 1919 | |||
| XM_011536926.1 | XP_011535228.1 | 13 | 1836 | |||
| GenBank mRNA | AB037754.1 | - | 14 | 1905 | ||
| AK000340.1 | - | 14 | 1918 | |||
| AK022867.1 | - | 4 | 728 | |||
| AK225346.1 | - | 14 | 1918 | |||
| AK296872.1 | - | 13 | 1877 | |||
| BC000973.2 | - | 14 | 1899 | 3450488 | ||
| BC040179.1 | - | 2 | 1209 | |||

G2E3
Genomic Map