siRNA Details
| Gene Symbol: G2E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308097.1 | NP_001295026.1 | 14 | 2427 | ||
| NM_017769.4 | NP_060239.2 | 15 | 2468 | |||
| XM_011536925.2 | XP_011535227.1 | 15 | 2434 | |||
| XM_011536926.1 | XP_011535228.1 | 14 | 2351 | |||
| GenBank mRNA | AB037754.1 | - | 15 | 2420 | ||
| AK000340.1 | - | 15 | 2433 | |||
| AK022867.1 | - | 5 | 1242 | |||
| AW963891.1 | - | 1 | 266 | |||
| BC000973.2 | - | 15 | 2414 | 3450488 | ||
| BC040179.1 | - | 3 | 1725 | |||

G2E3
Genomic Map