siRNA Details
Gene Symbol: WNT9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544271.2 | XP_011542573.1 | 4 | 1906 | ||
GenBank mRNA | AB060283.1 | - | 4 | 1345 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WNT9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544271.2 | XP_011542573.1 | 4 | 1906 | ||
GenBank mRNA | AB060283.1 | - | 4 | 1345 |