siRNA Details
| Gene Symbol: FLVCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195283.1 | NP_001182212.1 | 10 | 1185 | ||
| NM_017791.2 | NP_060261.2 | 10 | 2067 | |||
| GenBank mRNA | AK000378.1 | - | 10 | 2015 | ||
| AK027804.1 | - | 9 | 955 | |||
| AK297002.1 | - | 10 | 1185 | |||
| AY260572.1 | - | 10 | 2047 | |||
| AY260577.1 | - | 10 | 2047 | |||
| BC019087.2 | - | 10 | 2030 | 4866427 | ||

FLVCR2
Genomic Map