siRNA Details
| Gene Symbol: HAUS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166269.1 | NP_001159741.1 | 3 | 427 | ||
| NM_001166270.1 | NP_001159742.1 | 3 | 419 | |||
| NM_017815.2 | NP_060285.2 | 3 | 419 | |||
| GenBank mRNA | AK000431.1 | - | 3 | 388 | ||
| AK026016.1 | - | 3 | 373 | |||
| AK057779.1 | - | 3 | 495 | |||
| BC001916.1 | - | 3 | 416 | 2905474 | ||
| BC002554.2 | - | 3 | 350 | 3139198 | ||
| BQ066232.1 | - | 3 | 427 | |||
| BX247981.1 | - | 3 | 603 | |||
| BX248256.1 | - | 3 | 386 | |||

HAUS4
Genomic Map