siRNA Details
| Gene Symbol: GID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017896.2 | NP_060366.1 | 5 | 998 | ||
| GenBank mRNA | AK000609.1 | - | 5 | 934 | ||
| BC032120.2 | - | 5 | 968 | 4137113 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GID8| Gene Symbol: GID8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017896.2 | NP_060366.1 | 5 | 998 | ||
| GenBank mRNA | AK000609.1 | - | 5 | 934 | ||
| BC032120.2 | - | 5 | 968 | 4137113 | ||