siRNA Details
| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017924.3 | NP_060394.1 | 2 | 878 | ||
| XM_017021390.1 | XP_016876879.1 | 2 | 1052 | |||
| GenBank mRNA | AF061731.1 | - | 2 | 848 | ||
| AK000678.1 | - | 2 | 803 | |||
| BC009645.1 | - | 2 | 773 | 3458006 | ||
| BC056255.1 | - | 2 | 849 | 4694305 | ||
| BC061637.1 | - | 2 | 813 | 5532778 | ||

C14orf119
Genomic Map