siRNA Details
| Gene Symbol: FBXO34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017943.3 | NP_060413.2 | 2 | 3078 | ||
| XM_005267791.4 | XP_005267848.1 | 2 | 3267 | |||
| XM_006720185.3 | XP_006720248.1 | 2 | 2931 | |||
| XM_017021391.1 | XP_016876880.1 | 2 | 2995 | |||
| XM_017021392.1 | XP_016876881.1 | 2 | 4259 | |||
| XM_017021393.1 | XP_016876882.1 | 2 | 3362 | |||
| GenBank mRNA | AK000732.1 | - | 1 | 1621 | ||
| AK092873.1 | - | 1 | 2211 | |||
| AK092879.1 | - | 2 | 3020 | |||
| BC095482.1 | - | 2 | 2883 | 30334569 | ||
| BX248268.1 | - | 2 | 2930 | |||

FBXO34
Genomic Map