siRNA Details
| Gene Symbol: NRDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017970.3 | NP_060440.2 | 6 | 1578 | ||
| XM_011536886.1 | XP_011535188.1 | 6 | 1376 | |||
| XM_017021394.1 | XP_016876883.1 | 6 | 1376 | |||
| XM_017021395.1 | XP_016876884.1 | 7 | 1527 | |||
| GenBank mRNA | AK000870.1 | - | 6 | 1376 | ||
| AK024113.1 | - | 1 | 119 | |||
| AK294958.1 | - | 6 | 1382 | |||
| BC098568.1 | - | 6 | 1356 | 30706812 | ||
| BC118673.1 | - | 3 | 911 | 40028659 | ||

NRDE2
Genomic Map