siRNA Details
| Gene Symbol: AIM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039775.3 | NP_001034864.2 | 19 | 4987 | ||
| XM_005245918.2 | XP_005245975.1 | 19 | 4973 | |||
| XM_011541672.1 | XP_011539974.1 | 18 | 4937 | |||
| XM_011541673.2 | XP_011539975.1 | 19 | 5212 | |||
| GenBank mRNA | AK000902.1 | - | 3 | 1594 | ||
| AK095339.1 | - | 18 | 2031 | |||
| BC136870.1 | - | 15 | 1729 | 9020861 | ||

AIM1L
Genomic Map