siRNA Details
| Gene Symbol: GPATCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018025.2 | NP_060495.2 | 2 | 404 | ||
| NR_135270.1 | 2 | 404 | ||||
| XM_006723255.3 | XP_006723318.1 | 2 | 119 | |||
| GenBank mRNA | AF434677.1 | - | 2 | 90 | ||
| AK096005.1 | - | 2 | 98 | |||
| AK292503.1 | - | 2 | 98 | |||
| AK315712.1 | - | 2 | 116 | |||
| AL834465.1 | - | 2 | 95 | |||
| BC006108.1 | - | 2 | 404 | 3029084 | ||

GPATCH1
Genomic Map