siRNA Details
| Gene Symbol: ZNF205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042428.1 | NP_001035893.1 | 1986 | |||
| NM_001278158.1 | NP_001265087.1 | 2305 | ||||
| NM_003456.2 | NP_003447.2 | 2054 | ||||
| GenBank mRNA | AB672633.1 | - | 2298 | |||
| AF060865.1 | - | 2011 | ||||
| BC002810.2 | - | 2011 | 3635500 | |||
| BM924902.1 | - | 638 | ||||

ZNF205
Genomic Map