siRNA Details
Gene Symbol: BSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003458.3 | NP_003449.2 | 5 | 3980 | ||
GenBank mRNA | AF052224.1 | - | 5 | 3981 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003458.3 | NP_003449.2 | 5 | 3980 | ||
GenBank mRNA | AF052224.1 | - | 5 | 3981 |