siRNA Details
| Gene Symbol: LGI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018176.3 | NP_060646.2 | 6 | 733 | ||
| XM_011513850.2 | XP_011512152.1 | 6 | 635 | |||
| XM_017008356.1 | XP_016863845.1 | 6 | 635 | |||
| GenBank mRNA | AB067503.1 | - | 6 | 626 | ||
| AF467955.1 | - | 6 | 634 | |||
| AJ487516.1 | - | 6 | 634 | |||
| AJ487958.1 | - | 6 | 656 | |||
| AK126093.1 | - | 4 | 347 | |||
| AL833647.1 | - | 5 | 424 | |||
| BC101759.1 | - | 6 | 733 | 8069265 | ||
| BC101761.1 | - | 6 | 733 | 8069267 | ||

LGI2
Genomic Map