siRNA Details
| Gene Symbol: LRRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018214.4 | NP_060684.4 | 9 | 1101 | ||
| XM_011514726.2 | XP_011513028.1 | 10 | 1465 | |||
| XM_011514727.2 | XP_011513029.1 | 8 | 891 | |||
| XM_017010997.1 | XP_016866486.1 | 9 | 1393 | |||
| GenBank mRNA | AF332199.1 | - | 9 | 1048 | ||
| AF359380.1 | - | 9 | 824 | |||
| AK001637.1 | - | 9 | 804 | |||
| AK021896.1 | - | 10 | 1461 | |||
| AK292248.1 | - | 9 | 946 | |||
| BC003193.1 | - | 9 | 1101 | 3504090 | ||

LRRC1
Genomic Map