siRNA Details
| Gene Symbol: VRTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018228.2 | NP_060698.2 | 2 | 1496 | ||
| XM_011536911.1 | XP_011535213.1 | 3 | 1543 | |||
| GenBank mRNA | AK001673.1 | - | 2 | 1450 | ||
| AK023213.1 | - | 2 | 1463 | |||
| BC053325.1 | - | 2 | 1479 | 6577658 | ||

VRTN
Genomic Map