siRNA Details
| Gene Symbol: NUP133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018230.2 | NP_060700.2 | 8 | 1109 | ||
| GenBank mRNA | AK001676.1 | - | 8 | 1109 | ||
| AK092339.1 | - | 8 | 1066 | |||
| AK304061.1 | - | 9 | 1076 | |||
| AK304861.1 | - | 9 | 1010 | |||
| AK314431.1 | - | 8 | 1066 | |||
| BC020107.1 | - | 8 | 1093 | 3919973 | ||

NUP133
Genomic Map