siRNA Details
| Gene Symbol: KIRREL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286349.1 | NP_001273278.1 | 4 | 826 | ||
| NM_018240.6 | NP_060710.3 | 6 | 1126 | |||
| XM_005245305.4 | XP_005245362.1 | 6 | 783 | |||
| GenBank mRNA | AK022708.1 | - | 2 | 128 | ||
| AK297788.1 | - | 4 | 454 | |||
| AY017369.1 | - | 6 | 722 | |||
| AY302131.1 | - | 4 | 821 | |||
| BC109192.1 | - | 6 | 789 | 40001501 | ||
| BC109193.1 | - | 6 | 789 | 40001502 | ||

KIRREL
Genomic Map