siRNA Details
| Gene Symbol: ASXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018263.4 | NP_060733.4 | 4 | 425 | ||
| XM_011532950.2 | XP_011531252.1 | 3 | 464 | |||
| XM_011532951.2 | XP_011531253.1 | 2 | 152 | |||
| XM_017004429.1 | XP_016859918.1 | 4 | 255 | |||
| GenBank mRNA | AB051472.2 | - | 4 | 425 | ||
| AB084279.1 | - | 4 | 288 | |||
| BX647527.1 | - | 4 | 498 | |||
| BX648218.1 | - | 4 | 450 | |||
| CR933613.1 | - | 4 | 469 | |||

ASXL2
Genomic Map