siRNA Details
| Gene Symbol: MRGBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018270.5 | NP_060740.1 | 5 | 971 | ||
| NR_136405.1 | 5 | 1093 | ||||
| GenBank mRNA | AB085682.1 | - | 5 | 871 | ||
| AK001776.1 | - | 5 | 923 | |||
| BC009889.2 | - | 5 | 889 | 4026378 | ||
| BC014235.2 | - | 5 | 894 | 4555060 | ||
| BC018841.2 | - | 5 | 889 | 3050866 | ||

MRGBP
Genomic Map